Product Details

SNP ID
rs200927296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55090036 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGGAAATATGAGTGAGCTTCTA[A/G]GAGCAAGATCCCAATCCTCAGAAAG
Phenotype
MIM: 300738
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAGE2 PubMed Links

Gene Details

Gene
PAGE2
Gene Name
PAGE family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207339.3 76 Missense Mutation AGA,GGA R6G NP_997222.1
XM_017029353.1 76 Missense Mutation AGA,GGA R6G XP_016884842.1

View Full Product Details