Product Details

SNP ID
rs201527496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53082872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTCCCCACTCCTGAGGCCTCGG[G/T]GGGGAGCCTGGAAATCGATTTTCAG
Phenotype
MIM: 300253 MIM: 300564
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR173 PubMed Links

Gene Details

Gene
GPR173
Gene Name
G protein-coupled receptor 173
There are no transcripts associated with this gene.

Gene
TSPYL2
Gene Name
TSPY like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022117.3 546 Missense Mutation GGG,GTG G125V NP_071400.1
XM_006724592.3 546 Missense Mutation GGG,GTG G125V XP_006724655.1
XM_017029726.1 546 Missense Mutation GGG,GTG G125V XP_016885215.1
XM_017029727.1 546 Missense Mutation GGG,GTG G125V XP_016885216.1

View Full Product Details