Product Details

SNP ID
rs201634647
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48576552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAGTGGCAGGTATTATGACAGT[A/C]GACCTGGAGGGTATGGATATGGATA
Phenotype
MIM: 300027
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RBM3 PubMed Links

Gene Details

Gene
RBM3
Gene Name
RNA binding motif (RNP1, RRM) protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006743.4 595 Silent Mutation AGA,CGA R121R NP_006734.1

View Full Product Details