Product Details

SNP ID
rs200591905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49243161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTCCATGCCAGCAGGCTGGTCG[C/T]GCCAGAATTGAGTTCCAGAGGTGGT
Phenotype
MIM: 300110 MIM: 300859 MIM: 300292
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACNA1F PubMed Links

Gene Details

Gene
CACNA1F
Gene Name
calcium voltage-gated channel subunit alpha1 F
There are no transcripts associated with this gene.

Gene
CCDC22
Gene Name
coiled-coil domain containing 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014008.4 682 Missense Mutation GCG,GTG A171V NP_054727.1
XM_005272599.3 682 Missense Mutation GCG,GTG A171V XP_005272656.1
Gene
FOXP3
Gene Name
forkhead box P3
There are no transcripts associated with this gene.

View Full Product Details