Product Details

SNP ID
rs200960382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:107981533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCCTGGAAACACTGTAGCCTG[G/T]CCCTGTCCCTGGGGAGGTGCCAGCT
Phenotype
MIM: 300313
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105373310 PubMed Links

Gene Details

Gene
LOC105373310
Gene Name
NACHT, LRR and PYD domains-containing protein 10-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017030015.1 579 Intron XP_016885504.1
Gene
TEX13B
Gene Name
testis expressed 13B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031273.2 579 Silent Mutation GGA,GGC G162G NP_112563.1

View Full Product Details