Product Details

SNP ID
hCV192273577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:149884720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCATCAAAAATTACAAGAACCAC[C/T]TTCCTGATATCTTCAGCAAAGCCTC
Phenotype
MIM: 300892 MIM: 300341
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00850 PubMed Links

Gene Details

Gene
LINC00850
Gene Name
long intergenic non-protein coding RNA 850
There are no transcripts associated with this gene.

Gene
MAGEA8
Gene Name
MAGE family member A8
There are no transcripts associated with this gene.

Gene
MAGEA8-AS1
Gene Name
MAGEA8 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details