Product Details

SNP ID
rs201408833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154448722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATCGGCTTCGGGAAGAGCTGCG[A/G]CAGGAGTGGGAAGCCAAGCAGGAGA
Phenotype
MIM: 300453 MIM: 300104 MIM: 300022
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM50A PubMed Links

Gene Details

Gene
FAM50A
Gene Name
family with sequence similarity 50 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004699.3 662 Silent Mutation CGA,CGG R184R NP_004690.1
Gene
GDI1
Gene Name
GDP dissociation inhibitor 1
There are no transcripts associated with this gene.

Gene
MIR6858
Gene Name
microRNA 6858
There are no transcripts associated with this gene.

Gene
PLXNA3
Gene Name
plexin A3
There are no transcripts associated with this gene.

View Full Product Details