Product Details
- SNP ID
-
rs201174792
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:78657644 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGCCCTCTGACTCTCATGTAACT[A/T]TACAGCTGAACCAGGAACTCAGGAA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZCCHC5
PubMed Links
Gene Details
- Gene
- ZCCHC5
- Gene Name
- zinc finger CCHC-type containing 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152694.2 |
1073 |
Nonsense Mutation |
TAA,TAT |
*259Y |
NP_689907.1 |
View Full Product Details