Product Details

SNP ID
rs41279188
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.15:74720638 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
GCCAGGAAGAGAAAGACCTCCCAGC[G/T]GGCAATGGTCTCACCGATACACTTC
Phenotype
MIM: 108330
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
CYP1A1*5,g.2461C>A CYP1A1*5,g.2461C>T CYP1A1*9,g.2461C>T
Literature Links
CYP1A1 PubMed Links

Gene Details

Gene
CYP1A1
Gene Name
cytochrome P450 family 1 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000499.4 1589 Missense Mutation NP_000490.1
NM_001319216.1 1589 Missense Mutation NP_001306145.1
NM_001319217.1 1589 Missense Mutation NP_001306146.1
XM_017021953.1 1589 Missense Mutation XP_016877442.1

View Full Product Details