Product Details

SNP ID
rs9727
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150053482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCAACTTTTAGCTGCCACTTGG[C/G]TCATTACAGCAGTACCAGTATGGGG
Phenotype
MIM: 164770
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CSF1R PubMed Links

Gene Details

Gene
CSF1R
Gene Name
colony stimulating factor 1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288705.1 3798 UTR 3 NP_001275634.1
NM_005211.3 3798 UTR 3 NP_005202.2
Gene
HMGXB3
Gene Name
HMG-box containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014983.2 3798 Intron NP_055798.2
XM_006714765.2 3798 Intron XP_006714828.1

View Full Product Details