Product Details

SNP ID
rs10882649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95666329 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAGGCACTGAAAGAACCACATAC[A/G]TTACAAGGATGGCCAGCCTGGGCTG
Phenotype
MIM: 138250 MIM: 613847
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH18A1 PubMed Links

Gene Details

Gene
ALDH18A1
Gene Name
aldehyde dehydrogenase 18 family member A1
There are no transcripts associated with this gene.

Gene
TCTN3
Gene Name
tectonic family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143973.1 Intron NP_001137445.1
NM_015631.5 Intron NP_056446.4
XM_005269690.1 Intron XP_005269747.1
XM_011539627.1 Intron XP_011537929.1
XM_011539628.1 Intron XP_011537930.1

View Full Product Details