Product Details
- SNP ID
-
rs3746631
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:3668493 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGGAAGGTCCCCAAAATTATGTTTG[C/T]TTGCAGAGGCCAGCCAGGCTCCAGG
- Phenotype
-
MIM: 607114
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADAM33
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11905233] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADAM33
- Gene Name
- ADAM metallopeptidase domain 33
- Gene
- GFRA4
- Gene Name
- GDNF family receptor alpha 4
There are no transcripts associated with this gene.
View Full Product Details