Product Details

SNP ID
rs34665498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:287691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGACAAGCTGTGTTGAAGGCACT[C/T]GGTGGCGCGTACAATTGACAGAGGC
Phenotype
MIM: 602844 MIM: 603816 MIM: 608012
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGDIG PubMed Links

Gene Details

Gene
ARHGDIG
Gene Name
Rho GDP dissociation inhibitor gamma
There are no transcripts associated with this gene.

Gene
AXIN1
Gene Name
axin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003502.3 3420 UTR 3 NP_003493.1
NM_181050.2 3420 UTR 3 NP_851393.1
XM_011522682.2 3420 UTR 3 XP_011520984.1
XM_011522683.2 3420 UTR 3 XP_011520985.1
XM_011522684.2 3420 UTR 3 XP_011520986.1
XM_011522686.1 3420 UTR 3 XP_011520988.1
XM_017023743.1 3420 UTR 3 XP_016879232.1
XM_017023744.1 3420 UTR 3 XP_016879233.1
XM_017023745.1 3420 UTR 3 XP_016879234.1
XM_017023746.1 3420 UTR 3 XP_016879235.1
XM_017023747.1 3420 Intron XP_016879236.1
XM_017023748.1 3420 Intron XP_016879237.1
Gene
PDIA2
Gene Name
protein disulfide isomerase family A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006849.2 3420 Intron NP_006840.2

View Full Product Details