Product Details

SNP ID
rs34908430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:20784995 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTGAAGGCCGAGTGACTCAGGCC[C/T]GAGTAATCCGAGTTGTAGCCTCCGA
Phenotype
MIM: 608306
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SP8 PubMed Links

Gene Details

Gene
SP8
Gene Name
Sp8 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182700.5 1009 Silent Mutation NP_874359.2
NM_198956.3 1009 Silent Mutation NP_945194.1

View Full Product Details