Product Details
- SNP ID
-
rs3094972
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:22424068 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAATAAAATAAGTGAATTTATGGTA[C/T]GCAAGTTATACCTAAATAACATTGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
STEAP1B
PubMed Links
Gene Details
- Gene
- STEAP1B
- Gene Name
- STEAP family member 1B
View Full Product Details