Product Details

SNP ID
rs4500055
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:94824584 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTAATTGGCAGTGGTTATTTGCAGA[A/T]CCAGAAATCTATGACTGTTATGTTC
Phenotype
MIM: 611351
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
INTS8 PubMed Links

Gene Details

Gene
INTS8
Gene Name
integrator complex subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017864.3 Intron NP_060334.2
XM_017013616.1 Intron XP_016869105.1
XM_017013617.1 Intron XP_016869106.1
XM_017013618.1 Intron XP_016869107.1
XM_017013619.1 Intron XP_016869108.1

View Full Product Details