Product Details
- SNP ID
-
rs6850430
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:169736877 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCGATTTTAATTCTCGATGTTTAA[C/T]GTAGCAATAACTAGATTATGCGCAG
- Phenotype
-
MIM: 616614
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HPF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6832030] are located under a probe and SNP(s) [rs6850446] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HPF1
- Gene Name
- histone PARylation factor 1
View Full Product Details