Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001856.3 | Intron | NP_001847.3 | |||
XM_005270481.1 | Intron | XP_005270538.1 | |||
XM_011540722.1 | Intron | XP_011539024.1 | |||
XM_011540723.1 | Intron | XP_011539025.1 | |||
XM_011540724.1 | Intron | XP_011539026.1 | |||
XM_011540726.2 | Intron | XP_011539028.1 | |||
XM_011540727.2 | Intron | XP_011539029.1 | |||
XM_011540728.2 | Intron | XP_011539030.1 | |||
XM_011540729.1 | Intron | XP_011539031.1 | |||
XM_011540730.1 | Intron | XP_011539032.1 | |||
XM_017000338.1 | Intron | XP_016855827.1 | |||
XM_017000339.1 | Intron | XP_016855828.1 | |||
XM_017000340.1 | Intron | XP_016855829.1 | |||
XM_017000341.1 | Intron | XP_016855830.1 |