Product Details

SNP ID
rs12552646
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:100106193 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGTATGGTATCTACAAATCTACCT[C/T]GATCTGTACCCATCTTCTCTCTTCA
Phenotype
MIM: 609170 MIM: 243305
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERP44 PubMed Links

Gene Details

Gene
ERP44
Gene Name
endoplasmic reticulum protein 44
There are no transcripts associated with this gene.

Gene
INVS
Gene Name
inversin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318381.1 Intron NP_001305310.1
NM_001318382.1 Intron NP_001305311.1
NM_014425.4 Intron NP_055240.2

View Full Product Details