Product Details

SNP ID
rs7641637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:988744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCTGGTAGAGATGACTCTTATC[A/C]AGGAAAGAGGAACATACCTGGCCAA
Phenotype
MIM: 607220
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CNTN6 PubMed Links

Gene Details

Gene
CNTN6
Gene Name
contactin 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289080.1 Intron NP_001276009.1
NM_001289081.1 Intron NP_001276010.1
NM_014461.3 Intron NP_055276.1
XM_005265058.3 Intron XP_005265115.1
XM_011533590.2 Intron XP_011531892.1
XM_011533591.2 Intron XP_011531893.1
XM_017006171.1 Intron XP_016861660.1
XM_017006172.1 Intron XP_016861661.1
XM_017006173.1 Intron XP_016861662.1
XM_017006174.1 Intron XP_016861663.1
XM_017006175.1 Intron XP_016861664.1
XM_017006176.1 Intron XP_016861665.1
XM_017006177.1 Intron XP_016861666.1

View Full Product Details