Product Details
- SNP ID
-
rs2304277
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:9759396 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCCCTAAGCAGTTACTGTGTGCCCA[A/G]TGTGATGCCAGGTGCTGTGCAAGCT
- Phenotype
-
MIM: 604998
MIM: 601982
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CAMK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2069058] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CAMK1
- Gene Name
- calcium/calmodulin dependent protein kinase I
- Gene
- OGG1
- Gene Name
- 8-oxoguanine DNA glycosylase
View Full Product Details