Product Details

SNP ID
rs3766248
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:46307816 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TAGTTAAGCTCTGACCAAACAAGGG[A/T]TTTTAGGTTTAGCAGTTTTCATATT
Phenotype
MIM: 613844
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LRRC41 PubMed Links

Gene Details

Gene
LRRC41
Gene Name
leucine rich repeat containing 41
There are no transcripts associated with this gene.

Gene
UQCRH
Gene Name
ubiquinol-cytochrome c reductase hinge protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297565.1 Intron NP_001284494.1
NM_001297566.1 Intron NP_001284495.1
NM_006004.3 Intron NP_005995.2

View Full Product Details