Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001159488.1 | 1923 | UTR 3 | NP_001152960.1 | ||
NM_030764.3 | 1923 | UTR 3 | NP_110391.2 | ||
XM_006711535.3 | 1923 | Intron | XP_006711598.1 | ||
XM_011509974.2 | 1923 | UTR 3 | XP_011508276.1 | ||
XM_011509975.2 | 1923 | UTR 3 | XP_011508277.1 | ||
XM_011509976.2 | 1923 | UTR 3 | XP_011508278.1 | ||
XM_017002316.1 | 1923 | UTR 3 | XP_016857805.1 | ||
XM_017002317.1 | 1923 | UTR 3 | XP_016857806.1 | ||
XM_017002318.1 | 1923 | UTR 3 | XP_016857807.1 | ||
XM_017002319.1 | 1923 | UTR 3 | XP_016857808.1 |