Product Details

SNP ID
hCV11337019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47466199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGAAAAGCGACGAGGGCTCTCC[C/T]TGCCTTTTTTTAAACCATTAAAGTA
Phenotype
MIM: 601074
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELF1 PubMed Links

Gene Details

Gene
CELF1
Gene Name
CUGBP, Elav-like family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025596.2 4440 UTR 3 NP_001020767.1
NM_001172639.1 4440 UTR 3 NP_001166110.1
NM_001172640.1 4440 UTR 3 NP_001166111.1
NM_006560.3 4440 UTR 3 NP_006551.1
NM_198700.2 4440 UTR 3 NP_941989.1

View Full Product Details