Product Details

SNP ID
rs10533
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:154944469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCAACAGCACTTTATTGGGATCT[G/T]AGTCTACAGTTCACATAGGGAGGTG
Phenotype
MIM: 607622
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PBXIP1 PubMed Links

Gene Details

Gene
PBXIP1
Gene Name
PBX homeobox interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317734.1 2462 UTR 3 NP_001304663.1
NM_001317735.1 2462 UTR 3 NP_001304664.1
NM_020524.3 2462 UTR 3 NP_065385.2
Gene
PMVK
Gene Name
phosphomevalonate kinase
There are no transcripts associated with this gene.

View Full Product Details