Product Details

SNP ID
rs1034109
Assay Type
Functionally tested
NCBI dbSNP Submissions
71
Location
Chr.1:152787202 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGGACATGATGTTTCCATAATTA[C/T]AAGTCATGCTAAGAGTGTCAGAGGG
Phenotype
MIM: 612606 MIM: 612607
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCE1D PubMed Links

Gene Details

Gene
LCE1D
Gene Name
late cornified envelope 1D
There are no transcripts associated with this gene.

Gene
LCE1E
Gene Name
late cornified envelope 1E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178353.1 Intron NP_848130.1

View Full Product Details