Product Details

SNP ID
rs2834118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33170051 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAATACCCTGCTTCTTGTCCTA[A/G]GTTCCCAGGCTCTCTTTTTGCCTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01548 PubMed Links

Gene Details

Gene
LINC01548
Gene Name
long intergenic non-protein coding RNA 1548
There are no transcripts associated with this gene.

View Full Product Details