Product Details
- SNP ID
-
rs1995319
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:36290396 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGTATCTTGATGTGCTGAGTGATG[A/T]TACTGGCCCCCAAGTGTCTTGTTAT
- Phenotype
-
MIM: 616979
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DTHD1
PubMed Links
Gene Details
- Gene
- DTHD1
- Gene Name
- death domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001136536.4 |
1076 |
Missense Mutation |
GAT,GTT |
D14V |
NP_001130008.2 |
NM_001170700.2 |
1076 |
Missense Mutation |
GAT,GTT |
D179V |
NP_001164171.1 |
XM_006714014.3 |
1076 |
Missense Mutation |
GAT,GTT |
D304V |
XP_006714077.1 |
XM_011513693.2 |
1076 |
Missense Mutation |
GAT,GTT |
D304V |
XP_011511995.1 |
XM_011513694.2 |
1076 |
Missense Mutation |
GAT,GTT |
D304V |
XP_011511996.1 |
XM_011513695.2 |
1076 |
Missense Mutation |
GAT,GTT |
D304V |
XP_011511997.1 |
XM_011513696.2 |
1076 |
Missense Mutation |
GAT,GTT |
D14V |
XP_011511998.1 |
XM_017008191.1 |
1076 |
Missense Mutation |
GAT,GTT |
D304V |
XP_016863680.1 |
XM_017008192.1 |
1076 |
Missense Mutation |
GAT,GTT |
D304V |
XP_016863681.1 |
XM_017008193.1 |
1076 |
Missense Mutation |
GAT,GTT |
D304V |
XP_016863682.1 |
View Full Product Details