Product Details

SNP ID
rs1938916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66292830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTAGGTGTGCGCATGTGAGTGA[G/T]TCCCTTTGTGGGCAGGTCTCTCCCT
Phenotype
MIM: 611288 MIM: 611484
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CNIH2 PubMed Links

Gene Details

Gene
CNIH2
Gene Name
cornichon family AMPA receptor auxiliary protein 2
There are no transcripts associated with this gene.

Gene
TMEM151A
Gene Name
transmembrane protein 151A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153266.3 Intron NP_694998.1
Gene
YIF1A
Gene Name
Yip1 interacting factor homolog A, membrane trafficking protein
There are no transcripts associated with this gene.

View Full Product Details