Product Details

SNP ID
rs1939261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:89178067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGGAGAAGGAATGCTGTCCACC[G/A]TGGAGCGGGGACAGGAGTCCCTGTG
Phenotype
MIM: 606933
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
TYR PubMed Links

Gene Details

Gene
TYR
Gene Name
tyrosinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000372.4 503 Silent Mutation CCA,CCG P38P NP_000363.1
XM_011542970.2 503 Silent Mutation CCA,CCG P38P XP_011541272.1

View Full Product Details