Product Details
- SNP ID
-
rs2397
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:964887 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GAGCCTTTTAATAATGTGGCCAGCT[T/A]ATCATGACAAGCGTCTATGAGAAAT
- Phenotype
-
MIM: 608114
MIM: 610429
- Polymorphism
- T/A, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADAP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17227152] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADAP1
- Gene Name
- ArfGAP with dual PH domains 1
There are no transcripts associated with this gene.
- Gene
- COX19
- Gene Name
- COX19, cytochrome c oxidase assembly factor
View Full Product Details