Product Details

SNP ID
rs1874117
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:220791127 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCTGTGCTTCCTCCTGCCTTTGA[A/G]GTCTTCATGTAAAATAGATTCTTGT
Phenotype
MIM: 614126 MIM: 614127
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MARC1 PubMed Links

Gene Details

Gene
MARC1
Gene Name
mitochondrial amidoxime reducing component 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022746.3 Intron NP_073583.3
XM_011509900.2 Intron XP_011508202.1
XM_011509903.2 Intron XP_011508205.1
XM_011509904.2 Intron XP_011508206.1
XM_017002096.1 Intron XP_016857585.1
XM_017002097.1 Intron XP_016857586.1
Gene
MARC2
Gene Name
mitochondrial amidoxime reducing component 2
There are no transcripts associated with this gene.

View Full Product Details