Product Details

SNP ID
rs2836656
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:38747109 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGAAAGGTGGTCTCCAGAGCAA[A/G]GAAGCAGCACTAGTACCCCAGTTCA
Phenotype
MIM: 611723
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LINC00114 PubMed Links

Gene Details

Gene
LINC00114
Gene Name
long intergenic non-protein coding RNA 114
There are no transcripts associated with this gene.

View Full Product Details