Product Details

SNP ID
rs104894401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:20189154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAAGGCGGCTTCGAAGATGACC[T/C]GGAAGAAGATGCTGCTTGTGTAGGT
Phenotype
MIM: 121011
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
GJB2 PubMed Links

Gene Details

Gene
GJB2
Gene Name
gap junction protein beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004004.5 634 Missense Mutation CAG,CGG Q143R NP_003995.2
XM_011535049.2 634 Missense Mutation CAG,CGG Q143R XP_011533351.1

View Full Product Details