Product Details

SNP ID
rs2234687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:68154976 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTAAAGCACTGGCTCAGATTGCAG[A/G]CATATTTTCAAACCGGCAGTAACTG
Phenotype
MIM: 147570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFNG PubMed Links

Gene Details

Gene
IFNG
Gene Name
interferon gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000619.2 1004 UTR 3 NP_000610.2

View Full Product Details