Product Details

SNP ID
rs10234745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123457414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTGTTAGCTTGAGATGAGCAGC[C/T]GCTTCATCTTTGGTAAGAAGAATGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IQUB PubMed Links

Gene Details

Gene
IQUB
Gene Name
IQ motif and ubiquitin domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282855.1 2749 Silent Mutation GCA,GCG A720A NP_001269784.1
NM_001321293.1 2749 Silent Mutation GCA,GCG A720A NP_001308222.1
NM_178827.4 2749 Silent Mutation GCA,GCG A720A NP_849149.3
XM_005250161.3 2749 Silent Mutation GCA,GCG A720A XP_005250218.1
XM_005250162.4 2749 Silent Mutation GCA,GCG A720A XP_005250219.1
XM_011515833.2 2749 Silent Mutation GCA,GCG A720A XP_011514135.1
XM_011515834.2 2749 Silent Mutation GCA,GCG A720A XP_011514136.1
XM_017011771.1 2749 Silent Mutation GCA,GCG A720A XP_016867260.1
XM_017011772.1 2749 Silent Mutation GCA,GCG A441A XP_016867261.1

View Full Product Details