Product Details
- SNP ID
-
rs12548737
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:8703026 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGCAGCAGCGTCAGCACCATCCAA[A/G]TGGAGTGGCCCGAGCCCGACCTGGC
- Phenotype
-
MIM: 609203
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN23
PubMed Links
Gene Details
- Gene
- CLDN23
- Gene Name
- claudin 23
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_194284.2 |
871 |
Missense Mutation |
ATG,GTG |
M210V |
NP_919260.2 |
View Full Product Details