Product Details

SNP ID
rs1920148
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:171142645 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAGGAACTTGATAGAAATGCAAA[C/T]CACCCCAGACTCCACCCCAGACAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FMO6P PubMed Links

Gene Details

Gene
FMO6P
Gene Name
flavin containing monooxygenase 6 pseudogene
There are no transcripts associated with this gene.

View Full Product Details