Product Details

SNP ID
rs752150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57684839 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGTGATGTACATGAGAAGAGAGA[A/G]AATAAACTGAGGGCCATATGGGTGA
Phenotype
MIM: 613419
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF551 PubMed Links

Gene Details

Gene
ZNF551
Gene Name
zinc finger protein 551
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270938.1 Intron NP_001257867.1
NM_138347.4 Intron NP_612356.2
Gene
ZSCAN4
Gene Name
zinc finger and SCAN domain containing 4
There are no transcripts associated with this gene.

View Full Product Details