Product Details

SNP ID
rs12138571
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:19316179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTGGCTTTGTCATTGTGACGGT[A/G]GGGAGGTGGCTGTGGACTCCCTGCT
Phenotype
MIM: 603418 MIM: 614760
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKR7A2 PubMed Links

Gene Details

Gene
AKR7A2
Gene Name
aldo-keto reductase family 7 member A2
There are no transcripts associated with this gene.

Gene
PQLC2
Gene Name
PQ loop repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040125.1 Intron NP_001035214.1
NM_001040126.1 Intron NP_001035215.1
NM_001287531.1 Intron NP_001274460.1
NM_017765.2 Intron NP_060235.2
XM_005245916.2 Intron XP_005245973.1
XM_006710710.2 Intron XP_006710773.1
XM_006710711.2 Intron XP_006710774.1
XM_011541634.2 Intron XP_011539936.1

View Full Product Details