Product Details
- SNP ID
-
rs15610
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:112829945 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAAACTAGGCTCTTTTACAGACA[A/C]TGCTACTTCTTGCCCCCTTTGAATA
- Phenotype
-
MIM: 147720
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IL1B
PubMed Links
Gene Details
- Gene
- IL1B
- Gene Name
- interleukin 1 beta
View Full Product Details