Product Details

SNP ID
rs12130370
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:15484129 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTAGAGTTCAGAACAGCATTTCC[C/T]CTGTGACCTGAGGTCTCTGGAGCTC
Phenotype
MIM: 602234 MIM: 609444
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASP9 PubMed Links

Gene Details

Gene
CASP9
Gene Name
caspase 9
There are no transcripts associated with this gene.

Gene
CELA2B
Gene Name
chymotrypsin like elastase family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015849.2 Intron NP_056933.2

View Full Product Details