Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001297623.2 | 3379 | UTR 3 | NP_001284552.1 | ||
NM_001322821.1 | 3379 | UTR 3 | NP_001309750.1 | ||
NM_001322823.1 | 3379 | UTR 3 | NP_001309752.1 | ||
NM_001322825.1 | 3379 | UTR 3 | NP_001309754.1 | ||
NM_001322826.1 | 3379 | UTR 3 | NP_001309755.1 | ||
NM_001322827.1 | 3379 | UTR 3 | NP_001309756.1 | ||
NM_001322828.1 | 3379 | UTR 3 | NP_001309757.1 | ||
NM_153257.4 | 3379 | UTR 3 | NP_694989.2 | ||
XM_011527486.2 | 3379 | UTR 3 | XP_011525788.1 | ||
XM_017027470.1 | 3379 | UTR 3 | XP_016882959.1 |