Product Details

SNP ID
rs11592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219550252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCAAGGTCCCATGGGCCACCT[C/T]CTGCATGTCTCCGCGGAGGGGCTAC
Phenotype
MIM: 610405 MIM: 610991
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHPF PubMed Links

Gene Details

Gene
CHPF
Gene Name
chondroitin polymerizing factor
There are no transcripts associated with this gene.

Gene
MIR3132
Gene Name
microRNA 3132
There are no transcripts associated with this gene.

Gene
OBSL1
Gene Name
obscurin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173408.1 1892 Intron NP_001166879.1
NM_001173431.1 1892 Intron NP_001166902.1
NM_015311.2 1892 Intron NP_056126.1
XM_005246424.4 1892 Intron XP_005246481.1
XM_005246427.4 1892 Intron XP_005246484.1
XM_011510857.2 1892 Intron XP_011509159.1
XM_011510863.2 1892 Intron XP_011509165.1
XM_011510864.2 1892 Intron XP_011509166.1
XM_011510865.2 1892 Intron XP_011509167.1
XM_011510866.2 1892 Intron XP_011509168.1
XM_017003696.1 1892 Intron XP_016859185.1
XM_017003697.1 1892 Intron XP_016859186.1
XM_017003698.1 1892 Intron XP_016859187.1
XM_017003699.1 1892 Intron XP_016859188.1
XM_017003700.1 1892 Intron XP_016859189.1
Gene
TMEM198
Gene Name
transmembrane protein 198
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005209.2 1892 UTR 3 NP_001005209.1
NM_001303098.1 1892 UTR 3 NP_001290027.1
XM_017003367.1 1892 UTR 3 XP_016858856.1

View Full Product Details