Product Details

SNP ID
rs1815826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94574119 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAACATGCCAGAAAGTCAGCATC[C/T]CCCGTACTAATTTAAACAGAAAATA
Phenotype
MIM: 610315
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105369438 PubMed Links

Gene Details

Gene
LOC105369438
Gene Name
uncharacterized LOC105369438
There are no transcripts associated with this gene.

Gene
PIWIL4
Gene Name
piwi like RNA-mediated gene silencing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152431.2 Intron NP_689644.2

View Full Product Details