Product Details
- SNP ID
-
rs1902328
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:49632587 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCCAACCCTTCCCCCTCCTCTTT[C/T]TTCGGCAGACGGCTCCGGGTGCGGA
- Phenotype
-
MIM: 616288
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FMNL3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79252534] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FMNL3
- Gene Name
- formin like 3
There are no transcripts associated with this gene.
- Gene
- PRPF40B
- Gene Name
- pre-mRNA processing factor 40 homolog B
View Full Product Details