Product Details
- SNP ID
-
rs1939017
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:102712302 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGAAGTGGTGGTTCTCTTTTGGC[A/T]GCTGTCCTGGGAAGATGCCCAGTAG
- Phenotype
-
MIM: 120355
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MMP27
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114911002] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MMP27
- Gene Name
- matrix metallopeptidase 27
There are no transcripts associated with this gene.
- Gene
- MMP8
- Gene Name
- matrix metallopeptidase 8
View Full Product Details