Product Details

SNP ID
hCV11485560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38685695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAACGCAGCACAAGGCCAACAA[A/C]AAAAAGGAATAAAAGCAAGAAAAAC
Phenotype
MIM: 615323 MIM: 607046
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
JOSD1 PubMed Links

Gene Details

Gene
JOSD1
Gene Name
Josephin domain containing 1
There are no transcripts associated with this gene.

Gene
TOMM22
Gene Name
translocase of outer mitochondrial membrane 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020243.4 Intron NP_064628.1

View Full Product Details