Product Details

SNP ID
rs1946783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:103933022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGATCCCTATCAGTCAAGATAGG[A/C]TAGATTATTTTGCAGTGGCAAAAAA
Phenotype
MIM: 191175
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HSP90B1 PubMed Links

Gene Details

Gene
HSP90B1
Gene Name
heat shock protein 90 beta family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003299.2 Intron NP_003290.1
Gene
MIR3652
Gene Name
microRNA 3652
There are no transcripts associated with this gene.

Gene
TTC41P
Gene Name
tetratricopeptide repeat domain 41, pseudogene
There are no transcripts associated with this gene.

View Full Product Details