Product Details

SNP ID
rs2008188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:65964026 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATCCATGATGAGGTCCCGTTTTTC[G/C]TTCCTCCTTGGCTACAAAATAGACA
Phenotype
MIM: 611499 MIM: 608838
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
GUSB PubMed Links

Gene Details

Gene
GUSB
Gene Name
glucuronidase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000181.3 Intron NP_000172.2
NM_001284290.1 Intron NP_001271219.1
NM_001293104.1 Intron NP_001280033.1
NM_001293105.1 Intron NP_001280034.1
XM_005250297.3 Intron XP_005250354.1
XM_011516113.1 Intron XP_011514415.1
XM_011516114.1 Intron XP_011514416.1
XM_017012091.1 Intron XP_016867580.1
XM_017012092.1 Intron XP_016867581.1
XM_017012093.1 Intron XP_016867582.1
Gene
VKORC1L1
Gene Name
vitamin K epoxide reductase complex subunit 1 like 1
There are no transcripts associated with this gene.

View Full Product Details